Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- SMC1
- Entrez Gene ID
-
42853
- Gene Name
- Structural maintenance of chromosomes 1
- Gene Aliases
-
AAF56231, CG6057, DmSMC1, Dmel\CG6057, Dmel_CG6057, Smc1, dSMC1, smc1
- Chromosome Location
-
Chr.3R: 24058287 - 24062815 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0040283
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_142954.4 |
NP_651211.2 |
4 - 5 |
1849 |
76 |
View Full Product Details