Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
D19A
Entrez Gene ID
38718
Gene Name
CG10269-PA
Gene Aliases
CG10269, Dmel\CG10269, Dmel_CG10269, d19a
Chromosome Location
Chr.3L: 6185830 - 6189123 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0022935
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_057956.4 NP_477304.1 1 - 2 1090 117
GenBank mRNA BT016008.1 - 1 - 2 1087 117

View Full Product Details