Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- UQCR-14
- Entrez Gene ID
-
32586
- Gene Name
- Ubiquinol-cytochrome c reductase 14 kDa subunit
- Gene Aliases
-
BcDNA:RH44664, CG3560, Dmel\CG3560, Dmel_CG3560, UCR6, Ucrh, Ucrh 14KDa, anon-EST:Posey167
- Chromosome Location
-
Chr.X: 16286288 - 16286988 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0030733
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_132882.3 |
NP_573110.1 |
2 - 3 |
207 |
105 |
GenBank mRNA |
AY113594.1 |
- |
2 - 3 |
152 |
105 |
View Full Product Details