Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Obp47a
- Entrez Gene ID
-
36162
- Gene Name
- Odorant-binding protein 47a
- Gene Aliases
-
47a, CG12944, DmelObp47a, Dmel\CG12944, Dmel_CG12944
- Chromosome Location
-
Chr.2R: 10909923 - 10910485 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0033573
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_136788.1 |
NP_610632.1 |
1 - 2 |
53 |
100 |
View Full Product Details