Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- ProtB
- Entrez Gene ID
-
34867
- Gene Name
- Protamine B
- Gene Aliases
-
BG:DS03431.3, BcDNA:GH11850, CG4478, Dmel\CG4478, Dmel_CG4478, MST35Bb, Mst35Bb, dProtB, mst35b, prot-B, protB
- Chromosome Location
-
Chr.2L: 14882101 - 14883721 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0013301
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_057703.4 |
NP_477051.1 |
3 - 3 |
616 |
120 |
GenBank mRNA |
AY118776.1 |
- |
3 - 3 |
607 |
120 |
View Full Product Details