Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Sgs3
- Entrez Gene ID
-
39288
- Gene Name
- Salivary gland secretion 3
- Gene Aliases
-
68C GII, CG11720, DMSGS378, Dmel\CG11720, Dmel_CG11720, Glue, Muc68Cb, SGS-3, SGS378, Sgs, Sgs-3, sgs-3, sgs3
- Chromosome Location
-
Chr.3L: 11512221 - 11513402 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0003373
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_079300.3 |
NP_524024.1 |
2 - 2 |
945 |
74 |
View Full Product Details