Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Sdic1
Entrez Gene ID
43984
Gene Name
Sperm-specific dynein intermediate chain 1
Gene Aliases
CG9580, Dmel\CG9580, Dmel_CG9580, Sdic:CG9580
Chromosome Location
Chr.X: 20174217 - 20178638 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0067861
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001258848.1 NP_001245777.1 6 - 6 1640 115
NM_079931.4 NP_524670.2 5 - 5 1731 115

View Full Product Details