Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- SmD1
- Entrez Gene ID
-
44668
- Gene Name
- Small ribonucleoprotein particle protein SmD1
- Gene Aliases
-
BcDNA:RE39488, CG10753, D1-Sm, Dmel\CG10753, Dmel_CG10753, dD1, l(2)SH3 003, l(3)SH003, smd1, snRNP, snRNP-69D, snRNP69D
- Chromosome Location
-
Chr.3L: 12733364 - 12734611 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0261933
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_080035.3 |
NP_524774.1 |
2 - 2 |
675 |
78 |
GenBank mRNA |
BT032651.1 |
- |
2 - 2 |
672 |
78 |
View Full Product Details