Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Prosalpha1
Entrez Gene ID
45780
Gene Name
Proteasome alpha1 subunit
Gene Aliases
18495, Alpha-1, CG18495, Dmel\CG18495, Dmel_CG18495, PSA6_DROME, Pros alpha1, Prosalpha6, alpha-1, alpha1, l(2)SH2 2342, l(2)SH2342, prosalpha1, prosalpha6
Chromosome Location
Chr.2R: 7827303 - 7828678 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0263121
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_080098.4 NP_524837.2 4 - 4 858 62
NM_165565.3 NP_724614.1 4 - 4 791 62
GenBank mRNA AY118637.1 - 4 - 4 826 62

Gene Symbol
CG30382
Entrez Gene ID
246582
Gene Name
CG30382-PA
Gene Aliases
30382, Dmel\CG30382, Dmel_CG30382, Pros-alpha1, Prosalpha1R, Prosalpha6
Chromosome Location
Chr.2R: 7830413 - 7831771 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0050382
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001169607.1 NP_001163078.1 4 - 4 774 62
NM_165567.4 NP_724616.1 4 - 4 841 62

View Full Product Details