Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Tsp42Eb
Entrez Gene ID
59177
Gene Name
Tetraspanin 42Eb
Gene Aliases
BEST:LP06288, CG18816, Dm.Tsp42Eb, Dmel\CG18816, Dmel_CG18816, LP06288, Tetrasp42Eb
Chromosome Location
Chr.2R: 6998848 - 6999848 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0042086
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_165503.3 NP_724525.1 3 - 3 810 70
GenBank mRNA AF274015.1 - 3 - 3 789 70
AY060629.1 - 3 - 3 1493 70

Gene Symbol
CG30160
Entrez Gene ID
246490
Gene Name
CG30160-PA
Gene Aliases
Dmel\CG30160, Dmel_CG30160
Chromosome Location
Chr.2R: 7005157 - 7006136 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0050160
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_165504.2 NP_724526.1 3 - 3 789 70

View Full Product Details