Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- mtTFB1
- Entrez Gene ID
-
8674019
- Gene Name
- Mitochondrial Transcription Factor B1
- Gene Aliases
-
CG42631, CG7319, Dmel\CG42631, Dmel_CG42631, TFB1, d-TFB1M, d-mtTFB1
- Chromosome Location
-
Chr.3L: 11708979 - 11710570 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0261381
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_140242.4 |
NP_648499.1 |
2 - 2 |
889 |
75 |
- Gene Symbol
- Ccdc56
- Entrez Gene ID
-
39320
- Gene Name
- Coiled-coil domain containing 56
- Gene Aliases
-
CCDC56, CG42630, CG7319, Dmel\CG42630, Dmel_CG42630, ccdc56
- Chromosome Location
-
Chr.3L: 11708979 - 11710570 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0261353
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_206340.4 |
NP_996062.3 |
2 - 2 |
889 |
75 |
GenBank mRNA |
AY069635.1 |
- |
2 - 2 |
871 |
75 |
View Full Product Details