Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- spd-2
- Entrez Gene ID
-
39850
- Gene Name
- spindle defective 2
- Gene Aliases
-
BcDNA:LD24702, CEP192, CG17286, CG17286-PA, Cep192, D-SPD2, D-Spd2, DSPD-2, DSpd-2, DSpd2, Dmel\CG17286, Dmel_CG17286, Dspd-2, Dspd2, SPD-2, SPD2, Spd-2, Spd2, anon-73Ba, dSpd2, spd2
- Chromosome Location
-
Chr.3L: 16591207 - 16595014 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0027500
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_140649.4 |
NP_648906.1 |
3 - 3 |
3512 |
130 |
GenBank mRNA |
AF145687.1 |
- |
3 - 3 |
3466 |
130 |
View Full Product Details