Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Cpr97Eb
Entrez Gene ID
43259
Gene Name
Cuticular protein 97Eb
Gene Aliases
CG15884, CT33880, Dmel\CG15884, Dmel_CG15884
Chromosome Location
Chr.3R: 27089704 - 27091130 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0039481
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_143273.3 NP_651530.1 2 - 2 934 86
GenBank mRNA AY070959.1 - 2 - 2 927 86

View Full Product Details