Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Npc2e
- Entrez Gene ID
-
326136
- Gene Name
- Niemann-Pick type C-2e
- Gene Aliases
-
CG31410, CG3928, Dmel\CG31410, Dmel_CG31410, npc2e
- Chromosome Location
-
Chr.3R: 10053332 - 10054348 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0051410
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_169323.3 |
NP_731439.2 |
3 - 3 |
552 |
68 |
View Full Product Details