Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ssdp
Entrez Gene ID
42177
Gene Name
Sequence-specific single-stranded DNA-binding protein
Gene Aliases
CG7187, Dmel\CG7187, Dmel_CG7187, l(3)neo48, ssdp
Chromosome Location
Chr.3R: 18192494 - 18199595 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0011481
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001275769.1 NP_001262698.1 2 - 2 2775 73
NM_142431.1 NP_650688.1 2 - 2 2775 73
NM_169794.2 NP_732304.1 2 - 2 2270 73
NM_169795.1 NP_732305.1 2 - 2 2763 73

View Full Product Details