Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
p24-2
Entrez Gene ID
318890
Gene Name
p24-related-2
Gene Aliases
CG33105, CG9443, Dmel\CG33105, Dmel_CG33105, p24
Chromosome Location
Chr.3R: 9687402 - 9692794 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0053105
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001202276.2 NP_001189205.1 2 - 2 731 105
NM_176440.1 NP_788617.1 2 - 2 750 105
GenBank mRNA BT099592.1 - 2 - 2 709 105

Gene Symbol
Unc-115b
Entrez Gene ID
41178
Gene Name
Uncoordinated 115b
Gene Aliases
CG31332, CG9489, Dmel\CG31332, Dmel_CG31332, anon-WO0140519.9, unc-115
Chromosome Location
Chr.3R: 9687060 - 9692775 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0260463
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001202275.2 NP_001189204.1 2 - 2 731 105

View Full Product Details