Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
HP1b
Entrez Gene ID
31834
Gene Name
Heterochromatin Protein 1b
Gene Aliases
CG7041, Dmel\CG7041, Dmel_CG7041, HP1, HP1B, Hp1b, hp1b
Chromosome Location
Chr.X: 9081063 - 9082746 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0030082
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001169242.2 NP_001162713.1 1 - 1 820 69
NM_001258657.2 NP_001245586.1 1 - 1 820 69
NM_132293.3 NP_572521.2 2 - 2 925 69
GenBank mRNA BT100168.1 - 2 - 2 925 69
BT100169.1 - 2 - 2 925 69

View Full Product Details