Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- CCT6
- Entrez Gene ID
-
32518
- Gene Name
- Chaperonin containing TCP1 subunit 6
- Gene Aliases
-
CG8231, Cct6, Dmel\CG8231, Dmel_CG8231, L(1)g0022, Q9VXQ5_DROME, Rcd3, TCP-1-zeta, TCP-1zeta, TCP1-zeta, TCP1zeta, TCPzeta, Tcp-1-zeta, Tcp-1zeta, l(1)G0022, l(1)G0027, l(1)G0057
- Chromosome Location
-
Chr.X: 15746648 - 15748971 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0027329
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_132838.3 |
NP_573066.1 |
2 - 2 |
1806 |
84 |
GenBank mRNA |
AY051470.1 |
- |
2 - 2 |
1785 |
84 |
View Full Product Details