Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Cpr49Ah
- Entrez Gene ID
-
36354
- Gene Name
- Cuticular protein 49Ah
- Gene Aliases
-
CG8515, DmelCpr49Ah, Dmel\CG8515, Dmel_CG8515
- Chromosome Location
-
Chr.2R: 12413962 - 12415170 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0033731
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_136933.2 |
NP_610777.1 |
2 - 2 |
623 |
62 |
View Full Product Details