Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Nipped-B
Entrez Gene ID
3355136
Gene Name
CG17704-PE
Gene Aliases
CG17704, CG40281, Dmel\CG17704, Dmel_CG17704, NIPPED-B, NipB, l(2)02047, nipped-B
Chromosome Location
Chr.2R: 4691672 - 4729017 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0026401
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001042986.3 NP_001036451.1 2 - 3 333 77
NM_001042987.3 NP_001036452.1 2 - 3 361 77
NM_001169581.2 NP_001163052.1 2 - 3 326 77
NM_001169582.2 NP_001163053.1 2 - 3 361 77
NM_001169583.2 NP_001163054.1 2 - 3 333 77

View Full Product Details