Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PMCA
Entrez Gene ID
43787
Gene Name
plasma membrane calcium ATPase
Gene Aliases
BEST:CK, BEST:CK01140, BEST:CK02288, BP1021, CG2165, CG34036, CG42314, CK02288, Dmel\CG42314, Dmel_CG42314
Chromosome Location
Chr.4: 328193 - 358917 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0259214
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001038714.4 NP_001033803.3 17 - 18 4061 110

View Full Product Details