Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ppk10
Entrez Gene ID
3885617
Gene Name
pickpocket 10
Gene Aliases
BP1007, CG34042, Dmel\CG34042, Dmel_CG34042, PPK10
Chromosome Location
Chr.2L: 10450134 - 10452396 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0065110
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001038805.2 NP_001033894.1 3 - 4 519 86
GenBank mRNA AY226541.1 - 3 - 4 324 86

View Full Product Details