Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Muc96D
Entrez Gene ID
318737
Gene Name
Mucin 96D
Gene Aliases
CG31439, Dmel\CG31439, Dmel_CG31439
Chromosome Location
Chr.3R: 25397788 - 25400645 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0051439
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_170227.3 NP_733106.2 2 - 2 2554 61

View Full Product Details