Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ir21a
Entrez Gene ID
33157
Gene Name
Ionotropic receptor 21a
Gene Aliases
CG2657, CT8983, DmelIR21a, Dmel\CG2657, Dmel_CG2657, IR21a, ir21a
Chromosome Location
Chr.2L: 21823 - 25155 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0031209
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001103573.2 NP_001097043.1 2 - 3 695 102

View Full Product Details