Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Cpr65Ay
Entrez Gene ID
5740348
Gene Name
Cuticular protein 65Ay
Gene Aliases
CG34271, Dmel\CG34271, Dmel_CG34271
Chromosome Location
Chr.3L: 6158075 - 6158630 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0085300
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001104054.1 NP_001097524.1 2 - 3 162 84

View Full Product Details