Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
nvd
Entrez Gene ID
5740633
Gene Name
neverland
Gene Aliases
80Fc, CG40050, Dmel\CG40050, Dmel_CG40050, Nvd, l(3)80Fc, l(3)Lh6, l6, nvd-Dm
Chromosome Location
Chr.3L: 24324866 - 24401655 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0287185
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001104200.3 NP_001097670.1 4 - 5 1143 130
GenBank mRNA AB232987.1 - 4 - 5 953 130

View Full Product Details