Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
tau
Entrez Gene ID
326116
Gene Name
CG45110-PA
Gene Aliases
BcDNA:RE16764, CG12881, CG31057, CG31058, CG34294, CG45110, CG5606, Dmel\CG45110, Dmel_CG45110, Tau, d-tau, dTau, dtau
Chromosome Location
Chr.3R: 27639886 - 27656996 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0266579
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001300634.1 NP_001287563.1 7 - 7 1636 96
NM_001300635.1 NP_001287564.1 5 - 5 1291 96
NM_001300636.1 NP_001287565.1 6 - 6 1477 96

View Full Product Details