Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
Dbp80
Entrez Gene ID
3354919
Gene Name
Dead box protein 80
Gene Aliases
BcDNA:RE44177, CG17023, DBP80, DmDbp80, Dmel\CG17023, Dmel_CG17023, HEL80, Hel40, Hel80, dbp-80, dbp80, hel-40, mdcds_36124
Chromosome Location
Chr.3L: 27279316 - 27422159 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0024804
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001015151.3 NP_001015151.1 3 - 4 295 128
NM_001015152.4 NP_001015152.2 3 - 4 295 128
NM_001276239.1 NP_001263168.1 3 - 4 295 128
NM_001276241.1 NP_001263170.1 3 - 4 295 128

View Full Product Details