Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Mst57Dc
Entrez Gene ID
43121
Gene Name
Male-specific transcript 57Dc
Gene Aliases
Acp57Dc, CG4986, Dmel\CG4986, Dmel_CG4986, Mst75Dc, mst(2)ag-1, mst-1
Chromosome Location
Chr.3R: 25829321 - 25829698 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0011670
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_079774.4 NP_524498.2 1 - 1 210 65
GenBank mRNA DQ231598.1 - 1 - 1 196 65

View Full Product Details