Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
eIF4B
Entrez Gene ID
3355041
Gene Name
eukaryotic translation initiation factor 4B
Gene Aliases
CG10837, Dm-eIF4B, Dmel\CG10837, Dmel_CG10837, EIF-4B, Eif4B, eIF-4B, eIF-4B-L, eIF-4B-S, eIF-4b, eIF4B-L, eIF4B-S, eIf4b
Chromosome Location
Chr.3L: 27842669 - 27858271 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0020660
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001015320.2 NP_001015320.1 2 - 3 1231 132

View Full Product Details