Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
kl-5
Entrez Gene ID
26067054
Gene Name
male fertility factor kl5
Gene Aliases
CG12573, CG17616, CG40444, CG41497, CG45786, DHC4B, Dhc, Dhc-Yh3, DhcYh3, Dmel\CG45786, Dmel_CG45786, Kl-5, Lms4, kl5
Chromosome Location
Chr.Y: 22250 - 137486 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0267433
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001015499.4 NP_001015499.4 11 - 12 3579 112

View Full Product Details