Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
Ppr-Y
Entrez Gene ID
3355180
Gene Name
CG40440-PB
Gene Aliases
CG40440, Dmel\CG40440, Dmel_CG40440, PPr-Y, PprY
Chromosome Location
Chr.Y: 1636574 - 1884846 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0046697
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001015502.3 NP_001015502.3 5 - 6 1586 144
GenBank mRNA AF427495.1 - 5 - 6 1586 144

View Full Product Details