Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Snap25
Entrez Gene ID
3355084
Gene Name
Synaptosomal-associated protein 25kDa
Gene Aliases
CG17676, CG17884, CG40452, Dmel\CG40452, Dmel_CG40452, SNAP-25, SNAP25, dSNAP-25, snap-25, snap25, snp25
Chromosome Location
Chr.3L: 24074180 - 24299205 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0011288
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001043176.2 NP_001036641.1 3 - 4 274 62
GenBank mRNA BT023789.1 - 3 - 4 225 62
BT126217.1 - 3 - 4 226 62
L22021.1 - 3 - 4 223 62

View Full Product Details