Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
kdm5bb
Entrez Gene ID
415256
Gene Name
lysine (K)-specific demethylase 5Bb
Gene Aliases
cb264, jarid1bb, plu1, wu:fc44h11, zgc:85741
Chromosome Location
Chr.8: 28306539 - 28343108 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001002166.2 NP_001002166.2 14 - 15 2149 69
XM_005167024.4 XP_005167081.1 15 - 16 2827 69
XM_009303989.3 XP_009302264.1 15 - 16 2827 69
GenBank mRNA BC071280.1 - 14 - 15 2082 69

View Full Product Details