Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
trub2
Entrez Gene ID
431747
Gene Name
TruB pseudouridine (psi) synthase family member 2
Gene Aliases
fc51e05, wu:fc51e05, zgc:91836
Chromosome Location
Chr.6: 41023578 - 41028673 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001002200.2 NP_001002200.2 2 - 3 383 145
GenBank mRNA BC074078.1 - 2 - 3 356 145
EH452545.1 - 2 - 3 404 145
EH588883.1 - 2 - 3 415 145

View Full Product Details