Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
rab4b
Entrez Gene ID
445498
Gene Name
RAB4B, member RAS oncogene family
Gene Aliases
zgc:101015
Chromosome Location
Chr.15: 8813755 - 8856348 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001004002.1 NP_001004002.1 5 - 6 567 84
GenBank mRNA BC080219.1 - 5 - 6 567 84

View Full Product Details