Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
aspdh
Entrez Gene ID
447919
Gene Name
aspartate dehydrogenase domain containing
Gene Aliases
zgc:103722
Chromosome Location
Chr.3: 16747320 - 16762321 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001004657.1 NP_001004657.1 4 - 5 351 99
XM_005163809.4 XP_005163866.1 4 - 5 387 99
XM_021470416.1 XP_021326091.1 4 - 5 311 99
GenBank mRNA BC081519.1 - 4 - 5 351 99

View Full Product Details