Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
nitr1a
Entrez Gene ID
170994
Gene Name
novel immune-type receptor 1a
Gene Aliases
SI:dZ169I8.3, nitr1, nitr1.18, nitr1.19, nitr1.30, nitr1.60, nitr1.64, nitr1a_1, nitr1a_2, si:dz186f8.4
Chromosome Location
Chr.7: 235287 - 237232 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001007219.1 NP_001007220.1 4 - 5 870 91

View Full Product Details