Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
lrrc20
Entrez Gene ID
492485
Gene Name
leucine rich repeat containing 20
Gene Aliases
zgc:103699
Chromosome Location
Chr.12: 48214556 - 48240134 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001007357.1 NP_001007358.1 4 - 5 586 59
XM_005172993.4 XP_005173050.2 4 - 5 554 59
XM_005172994.4 XP_005173051.2 4 - 5 646 59
XM_005172995.4 XP_005173052.2 4 - 5 637 59
XM_005172996.4 XP_005173053.2 4 - 5 568 59
XM_021480056.1 XP_021335731.1 4 - 5 583 59
GenBank mRNA BC085589.1 - 4 - 5 586 59

View Full Product Details