Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- rpe65b
- Entrez Gene ID
-
100002865
- Gene Name
- retinoid isomerohydrolase RPE65 b
- Gene Aliases
-
RPE56b, rpepb, si:ch211-198n5.5, wu:fc74c09, zgc:110538
- Chromosome Location
-
Chr.8: 41182 - 50728 on Build GRCz11
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
BC092162.1 |
- |
8 - 9 |
888 |
129 |
View Full Product Details