Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
prrg4
Entrez Gene ID
550330
Gene Name
proline rich Gla (G-carboxyglutamic acid) 4 (transmembrane)
Gene Aliases
zgc:110234
Chromosome Location
Chr.18: 45666665 - 45674932 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001017637.2 NP_001017637.1 3 - 4 433 75
GenBank mRNA BC092813.1 - 3 - 4 374 75
EH538457.1 - 3 - 4 456 75

View Full Product Details