The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001024397.1 | NP_001019568.1 | 2 - 3 | 282 | 89 |
GenBank mRNA | BC095257.1 | - | 2 - 3 | 282 | 89 |
BC165709.1 | - | 2 - 3 | 297 | 89 |