Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
nsfb
Entrez Gene ID
554200
Gene Name
N-ethylmaleimide-sensitive factor b
Gene Aliases
-
Chromosome Location
Chr.12: 248424 - 299403 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001024454.2 NP_001019625.2 19 - 20 2215 91
NM_001354163.1 NP_001341092.1 19 - 20 2215 91
XM_005156207.4 XP_005156264.1 19 - 20 2252 91
XM_017358461.2 XP_017213950.1 18 - 19 2062 91
GenBank mRNA BC162793.1 - 19 - 20 2193 91
BC162811.1 - 19 - 20 2193 91
DQ022212.1 - 19 - 20 2215 91

View Full Product Details