Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
nsfb
Entrez Gene ID
554200
Gene Name
N-ethylmaleimide-sensitive factor b
Gene Aliases
-
Chromosome Location
Chr.12: 248424 - 299403 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001024454.2 NP_001019625.2 20 - 21 2267 99
NM_001354163.1 NP_001341092.1 20 - 21 2267 99
XM_005156207.4 XP_005156264.1 20 - 21 2304 99
XM_017358461.2 XP_017213950.1 19 - 20 2114 99
GenBank mRNA BC162793.1 - 20 - 21 99
BC162811.1 - 20 - 21 99
DQ022212.1 - 20 - 21 2267 99

View Full Product Details