Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
nsfb
Entrez Gene ID
554200
Gene Name
N-ethylmaleimide-sensitive factor b
Gene Aliases
-
Chromosome Location
Chr.12: 248424 - 299403 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA BC162793.1 - 2 - 3 135 99
BC162811.1 - 2 - 3 135 99
DQ022212.1 - 2 - 3 157 99

View Full Product Details