Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
fynb
Entrez Gene ID
574422
Gene Name
FYN proto-oncogene, Src family tyrosine kinase b
Gene Aliases
im:6908851, si:dkey-119m7.5, zgc:109996
Chromosome Location
Chr.20: 328942 - 397237 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001029969.1 NP_001025140.1 12 - 13 1632 124
XM_005160302.4 XP_005160359.1 12 - 13 1679 124
XM_009294487.3 XP_009292762.1 11 - 12 1514 124
XM_021468512.1 XP_021324187.1 11 - 12 1621 124
GenBank mRNA BC098534.1 - 12 - 13 1632 124

View Full Product Details