Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
kctd8
Entrez Gene ID
568933
Gene Name
potassium channel tetramerization domain containing 8
Gene Aliases
dex
Chromosome Location
Chr.14: 5936996 - 5938068 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001037241.1 NP_001032318.1 1 - 1 653 80
GenBank mRNA AY763410.1 - 1 - 1 653 80
BC162914.1 - 1 - 1 561 80
BC162947.1 - 1 - 1 561 80

View Full Product Details