Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
pglyrp5
Entrez Gene ID
553387
Gene Name
peptidoglycan recognition protein 5
Gene Aliases
PGRP-SC1a, si:ch211-202c21.3, si:dkey-113g17.4
Chromosome Location
Chr.18: 125195 - 128460 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001044321.1 NP_001037786.1 3 - 4 524 89
GenBank mRNA BC093314.1 - 3 - 4 407 89
DQ447203.1 - 3 - 4 524 89

View Full Product Details