Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
grm6b
Entrez Gene ID
565450
Gene Name
glutamate receptor, metabotropic 6b
Gene Aliases
cb496, grma, zK13A21.16, zgc:158329
Chromosome Location
Chr.11: 248793 - 292984 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001080020.1 NP_001073489.1 11 - 12 3019 141
GenBank mRNA BC129205.1 - 11 - 12 3019 141

View Full Product Details