Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
elna
Entrez Gene ID
790915
Gene Name
elastin a
Gene Aliases
eln1
Chromosome Location
Chr.15: 263189 - 323096 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001080063.2 NP_001073532.2 34 - 35 2352 97
XM_009291337.3 XP_009289612.1 34 - 35 2618 97
XM_017351021.2 XP_017206510.1 34 - 35 2619 97
XM_017351022.2 XP_017206511.1 34 - 35 2614 97
XM_017351023.2 XP_017206512.1 34 - 35 2614 97
XM_017351024.2 XP_017206513.1 34 - 35 2614 97
XM_017351025.2 XP_017206514.1 34 - 35 2619 97
XM_017351026.2 XP_017206515.1 34 - 35 2604 97
XM_017351027.2 XP_017206516.1 34 - 35 2619 97
XM_017351028.2 XP_017206517.1 34 - 35 2618 97
XM_017351029.2 XP_017206518.1 33 - 34 2574 97
XM_017351031.2 XP_017206520.1 32 - 33 2491 97
XM_017351032.2 XP_017206521.1 32 - 33 2470 97
XM_017351033.2 XP_017206522.1 30 - 31 2340 97
XM_017351035.2 XP_017206524.1 30 - 31 2341 97
XM_017351036.2 XP_017206525.1 30 - 31 2341 97
XM_017351037.2 XP_017206526.1 30 - 31 2341 97
XM_017351038.2 XP_017206527.1 26 - 27 2031 97
XM_021481472.1 XP_021337147.1 31 - 32 2577 97
XM_021481473.1 XP_021337148.1 28 - 29 2202 97
XM_021481474.1 XP_021337149.1 23 - 24 1989 97
GenBank mRNA DQ523562.1 - 34 - 35 2352 97
MF039671.1 - 11 - 12 816 97
MF039672.1 - 11 - 12 816 97
MF039673.1 - 11 - 12 816 97
MF039674.1 - 11 - 12 97
MF039675.1 - - 97
MF039676.1 - 5 - 6 318 97
MF039677.1 - 5 - 6 97
MF039678.1 - 5 - 6 318 97
MF101833.1 - 5 - 6 97

View Full Product Details